Product Details

SNP ID
rs146829849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32974128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTGAACAATACTTCCCAGCCCT[A/G]CAGAAGTTTACTGTGCTGGACCTTG
Phenotype
MIM: 615470 MIM: 610884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP89 PubMed Links

Gene Details

Gene
CEP89
Gene Name
centrosomal protein 89
There are no transcripts associated with this gene.

Gene
FAAP24
Gene Name
Fanconi anemia core complex associated protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300978.1 Intron NP_001287907.1
NM_152266.4 Intron NP_689479.1
XM_005259393.3 Intron XP_005259450.1
Gene
RHPN2
Gene Name
rhophilin Rho GTPase binding protein 2
There are no transcripts associated with this gene.

View Full Product Details