Product Details

SNP ID
rs147224039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44810753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGTTGGGATAAGCAAAGGCTG[A/G]TGCAGTATTACCCCAGGAGTGTTCA
Phenotype
MIM: 612773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAM PubMed Links

Gene Details

Gene
BCAM
Gene Name
basal cell adhesion molecule (Lutheran blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013257.2 Intron NP_001013275.1
NM_005581.4 Intron NP_005572.2

View Full Product Details