Product Details

SNP ID
rs147658534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8586687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTGGGGGGTCCCAGGCAGCCC[C/G]TCCAGCAGCAGGGACTCCTGGTCTC
Phenotype
MIM: 608990 MIM: 601480
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS10 PubMed Links

Gene Details

Gene
ADAMTS10
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282352.1 2580 Missense Mutation GAC,GAG D245E NP_001269281.1
NM_030957.3 2580 Missense Mutation GAC,GAG D758E NP_112219.3
XM_006722917.3 2580 Missense Mutation GAC,GAG D390E XP_006722980.1
XM_017027338.1 2580 Missense Mutation GAC,GAG D758E XP_016882827.1
XM_017027339.1 2580 Missense Mutation GAC,GAG D281E XP_016882828.1
XM_017027340.1 2580 Missense Mutation GAC,GAG D279E XP_016882829.1
Gene
MYO1F
Gene Name
myosin IF
There are no transcripts associated with this gene.

View Full Product Details