Product Details

SNP ID
rs147803651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57537341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTGAGATGTGTATCCTGGTC[A/T]TGAAAGACATTTTGTACCTCAGTGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199295.1 661 Missense Mutation ATG,TTG M113L NP_001186224.1
NM_153263.2 661 Missense Mutation ATG,TTG M100L NP_694995.2
XM_006723128.3 661 Missense Mutation ATG,TTG M91L XP_006723191.1
Gene
ZNF550
Gene Name
zinc finger protein 550
There are no transcripts associated with this gene.

View Full Product Details