Product Details

SNP ID
rs147939830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55489907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGCGTGTGGCTAACTCCAGGGA[C/T]GACTCAACATCTCCCCTGGATGGGG
Phenotype
MIM: 610671
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAT14 PubMed Links

Gene Details

Gene
NAT14
Gene Name
N-acetyltransferase 14 (putative)
There are no transcripts associated with this gene.

Gene
SSC5D
Gene Name
scavenger receptor cysteine rich family member with 5 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144950.1 474 Silent Mutation GAC,GAT D129D NP_001138422.1
NM_001195267.1 474 Silent Mutation GAC,GAT D129D NP_001182196.1
Gene
ZNF628
Gene Name
zinc finger protein 628
There are no transcripts associated with this gene.

View Full Product Details