Product Details

SNP ID
rs148042819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15807151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCATGGCGACCTTCAGCTCC[C/T]TGTTCCTGAGGCTGAAGATGATGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10H1 PubMed Links

Gene Details

Gene
OR10H1
Gene Name
olfactory receptor family 10 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013940.2 976 Missense Mutation AAG,AGG K296R NP_039228.1
XM_011527911.1 976 Missense Mutation AAG,AGG K296R XP_011526213.1

View Full Product Details