Product Details

SNP ID
rs148649128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1605373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGTCCTCACCAGTTCTTGACCA[C/G]CTCCCGGTAGCGTGGGCCCAGGAAC
Phenotype
MIM: 147141 MIM: 609711
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCF3 PubMed Links

Gene Details

Gene
TCF3
Gene Name
transcription factor 3
There are no transcripts associated with this gene.

Gene
UQCR11
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit XI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006830.3 112 Missense Mutation CTG,GTG L13V NP_006821.1

View Full Product Details