Product Details

SNP ID
rs148710691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55544923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTGGTCGTCAGCTCCACCAG[C/G]CGTTGGAGGGCTGTGGCTGTGTCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
There are no transcripts associated with this gene.

Gene
SBK3
Gene Name
SH3 domain binding kinase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199824.2 72 Silent Mutation CGC,CGG R24R NP_001186753.1
XM_011526298.2 72 Silent Mutation CGC,CGG R49R XP_011524600.1

View Full Product Details