Product Details

SNP ID
rs149721174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8302802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCTGCAATAACCCCATAGGC[A/G]GTTGGGCTTCCAGACTGGTCTCCGG
Phenotype
MIM: 606475 MIM: 602139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD320 PubMed Links

Gene Details

Gene
CD320
Gene Name
CD320 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165895.1 747 Silent Mutation ACC,ACT T185T NP_001159367.1
NM_016579.3 747 Silent Mutation ACC,ACT T227T NP_057663.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.

View Full Product Details