Product Details

SNP ID
rs149818710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53994769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGGATGTCCCAAGCCCTCGAT[A/G]GGACAGTGGGATGCCAGGCCCCCCG
Phenotype
MIM: 606898 MIM: 606900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG6 PubMed Links

Gene Details

Gene
CACNG6
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031897.2 Intron NP_114103.2
NM_145814.1 Intron NP_665813.1
NM_145815.1 Intron NP_665814.1
XM_006723316.3 Intron XP_006723379.1
Gene
CACNG8
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
There are no transcripts associated with this gene.

View Full Product Details