Product Details

SNP ID
rs150175128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:650260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCCCAGGCTGGGGATGGTGGC[A/G]GGCGTGATGATGCCGTTGACGAGCT
Phenotype
MIM: 605831 MIM: 615177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF22 PubMed Links

Gene Details

Gene
FGF22
Gene Name
fibroblast growth factor 22
There are no transcripts associated with this gene.

Gene
RNF126
Gene Name
ring finger protein 126
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194460.2 591 Intron NP_919442.1
XM_005259594.1 591 Silent Mutation CCC,CCT P133P XP_005259651.1

View Full Product Details