Product Details

SNP ID
rs111943844
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:149923838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGGAGGTGGCTAGAGCCGACT[G/T]ACCCATGGGAGATCCGAATGGAGGC
Phenotype
MIM: 605593 MIM: 185860
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTMR11 PubMed Links

Gene Details

Gene
MTMR11
Gene Name
myotubularin related protein 11
There are no transcripts associated with this gene.

Gene
SF3B4
Gene Name
splicing factor 3b subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005850.4 Intron NP_005841.1
Gene
SV2A
Gene Name
synaptic vesicle glycoprotein 2A
There are no transcripts associated with this gene.

View Full Product Details