Product Details

SNP ID
rs113343719
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:182900089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTGAAGAGCATTTTATCATCT[G/T]TTTTATCTCCTGAAGAAGCCCTTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 2141 Missense Mutation AAA,ACA K619T NP_112195.2

View Full Product Details