Product Details

SNP ID
rs113596156
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:1179319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAAGGATCCAGCAGAGGCCTC[A/G]GGCTCGAGTCTCAGGTGAATAGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTLL10 PubMed Links

Gene Details

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130045.1 414 Missense Mutation CAG,CGG Q35R NP_001123517.1
NM_153254.2 414 Intron NP_694986.2
XM_005244738.1 414 Intron XP_005244795.1
XM_011541177.2 414 Missense Mutation CAG,CGG Q35R XP_011539479.1
XM_017000906.1 414 Missense Mutation CAG,CGG Q35R XP_016856395.1
XM_017000907.1 414 Missense Mutation CAG,CGG Q35R XP_016856396.1
XM_017000908.1 414 Missense Mutation CAG,CGG Q35R XP_016856397.1
XM_017000909.1 414 Missense Mutation CAG,CGG Q35R XP_016856398.1
XM_017000910.1 414 Missense Mutation CAG,CGG Q35R XP_016856399.1
XM_017000911.1 414 Missense Mutation CAG,CGG Q35R XP_016856400.1
XM_017000912.1 414 Missense Mutation CAG,CGG Q35R XP_016856401.1
Gene
TTLL10-AS1
Gene Name
TTLL10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details