Product Details
- SNP ID
-
rs115217066
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:52057888 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTTGATTTGAAGAGTTAATTTT[A/G]TTCTTGCTTTTATTTCAAACAAGTC
- Phenotype
-
MIM: 609448
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTF3L4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201014028] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTF3L4
- Gene Name
- basic transcription factor 3 like 4
- Gene
- TXNDC12
- Gene Name
- thioredoxin domain containing 12
There are no transcripts associated with this gene.
- Gene
- TXNDC12-AS1
- Gene Name
- TXNDC12 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details