Product Details

SNP ID
rs116010861
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.1:24900495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTCTAAGCCTTTCTAGGGCCA[A/G]AGAACTCTGATGTGAGAATCCATGC
Phenotype
MIM: 600210
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RUNX3 PubMed Links

Gene Details

Gene
RUNX3
Gene Name
runt related transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031680.2 3286 UTR 3 NP_001026850.1
NM_001320672.1 3286 UTR 3 NP_001307601.1
NM_004350.2 3286 UTR 3 NP_004341.1
XM_005246024.4 3286 UTR 3 XP_005246081.1
XM_011542351.1 3286 UTR 3 XP_011540653.1
XM_017002670.1 3286 Intron XP_016858159.1

View Full Product Details