Product Details

SNP ID
rs116141493
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:222824900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTACAACCACAAGTTATTGCCAG[A/C]TGAAAAGGATAATGGAGGGTAATGA
Phenotype
MIM: 607502
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DISP1 PubMed Links

Gene Details

Gene
DISP1
Gene Name
dispatched RND transporter family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032890.3 Intron NP_116279.2
XM_005273335.2 Intron XP_005273392.1
XM_006711592.2 Intron XP_006711655.1
XM_006711594.3 Intron XP_006711657.1
XM_011510072.2 Intron XP_011508374.1
XM_011510073.2 Intron XP_011508375.1
XM_011510074.2 Intron XP_011508376.1
XM_011510075.2 Intron XP_011508377.1
XM_011510077.2 Intron XP_011508379.1
XM_017002611.1 Intron XP_016858100.1
XM_017002612.1 Intron XP_016858101.1
XM_017002613.1 Intron XP_016858102.1

View Full Product Details