Product Details

SNP ID
rs116317151
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:172537463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTGTGCTTGCATTATTAAATT[A/C]TTTGCAGTAATTTTTTTAAAAGCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 Intron NP_001269679.1
NM_001282751.1 Intron NP_001269680.1
NM_014283.4 Intron NP_055098.1
NM_016227.3 Intron NP_057311.3
XM_006711374.2 Intron XP_006711437.1
XM_006711375.2 Intron XP_006711438.1
XM_006711376.1 Intron XP_006711439.1

View Full Product Details