Product Details

SNP ID
rs116467545
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:196227600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTTATGTTTTAGTTTCTTCA[A/G]TGTAATAAAACATGCTATGAACATC
Phenotype
MIM: 610044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNT2 PubMed Links

Gene Details

Gene
KCNT2
Gene Name
potassium sodium-activated channel subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287819.1 3164 UTR 3 NP_001274748.1
NM_001287820.1 3164 UTR 3 NP_001274749.1
NM_198503.3 3164 UTR 3 NP_940905.2
XM_006711294.2 3164 UTR 3 XP_006711357.1
XM_006711295.2 3164 UTR 3 XP_006711358.1
XM_011509482.2 3164 UTR 3 XP_011507784.1
XM_011509483.2 3164 Intron XP_011507785.1
XM_017001178.1 3164 Intron XP_016856667.1
XM_017001179.1 3164 Intron XP_016856668.1
XM_017001180.1 3164 Intron XP_016856669.1
XM_017001181.1 3164 UTR 3 XP_016856670.1
XM_017001182.1 3164 UTR 3 XP_016856671.1
XM_017001183.1 3164 UTR 3 XP_016856672.1
XM_017001184.1 3164 UTR 3 XP_016856673.1
XM_017001185.1 3164 Intron XP_016856674.1

View Full Product Details