Product Details

SNP ID
rs138035892
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:233906046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACGGAGGACACTCCTATGTGGCA[G/T]ACCGTGGGAAACAGATCAGAAACTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC35F3 PubMed Links

Gene Details

Gene
SLC35F3
Gene Name
solute carrier family 35 member F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300845.1 Intron NP_001287774.1
NM_173508.3 Intron NP_775779.1

View Full Product Details