Product Details

SNP ID
rs138410930
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:35888452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGGCGCTTACCTGCCCCCCCT[C/G]CAGCAGGTGTTCCAGGCACCTCGCC
Phenotype
MIM: 606228
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317122.1 239 Silent Mutation CTC,CTG L17L NP_001304051.1
NM_001317123.1 239 UTR 5 NP_001304052.1
NM_012199.4 239 Silent Mutation CTC,CTG L17L NP_036331.1
XM_011541236.2 239 Silent Mutation CTC,CTG L17L XP_011539538.1
XM_017001016.1 239 Silent Mutation CTC,CTG L17L XP_016856505.1
XM_017001017.1 239 Silent Mutation CTC,CTG L17L XP_016856506.1
XM_017001018.1 239 Intron XP_016856507.1
XM_017001019.1 239 UTR 5 XP_016856508.1

View Full Product Details