Product Details

SNP ID
rs139132904
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:108715306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTGACTGACCAGAGTGGTTG[A/G]TTGCACATCTAGGGAATAAAGTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 Intron NP_001138409.1

View Full Product Details