Product Details

SNP ID
rs139426438
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:160216322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGTGTATGTATGTGTGTGGTGT[C/G]TGTGTACATGTCTTCTGTCACAAAG
Phenotype
MIM: 615820 MIM: 603434
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF8 PubMed Links

Gene Details

Gene
DCAF8
Gene Name
DDB1 and CUL4 associated factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015726.3 3276 UTR 3 NP_056541.2
Gene
LOC729867
Gene Name
uncharacterized LOC729867
There are no transcripts associated with this gene.

Gene
PEA15
Gene Name
phosphoprotein enriched in astrocytes 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297576.1 3276 Intron NP_001284505.1
NM_001297577.1 3276 Intron NP_001284506.1
NM_001297578.1 3276 Intron NP_001284507.1
NM_003768.4 3276 Intron NP_003759.1
XM_006711599.2 3276 Intron XP_006711662.1

View Full Product Details