Product Details

SNP ID
rs139452026
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:116375404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGCCATGGAGATGGTTTGGGTA[A/C]CTAAGTTATGTCCATTTATCTAGAT
Phenotype
MIM: 182310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A1 PubMed Links

Gene Details

Gene
ATP1A1
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000701.7 Intron NP_000692.2
NM_001160233.1 Intron NP_001153705.1
NM_001160234.1 Intron NP_001153706.1
XM_017001360.1 Intron XP_016856849.1
XM_017001361.1 Intron XP_016856850.1

View Full Product Details