Product Details

SNP ID
rs140192316
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:59821916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCAAACAGCTGACTAGTGGAG[C/T]TGCCATGGCACAAGTTCTTCATCAA
Phenotype
MIM: 607820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOOK1 PubMed Links

Gene Details

Gene
HOOK1
Gene Name
hook microtubule-tethering protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015888.4 318 Missense Mutation GCT,GTT A41V NP_056972.1
XM_006710676.1 318 Missense Mutation GCT,GTT A41V XP_006710739.1
XM_011541562.2 318 UTR 5 XP_011539864.1
XM_011541563.1 318 Missense Mutation GCT,GTT A41V XP_011539865.1
XM_017001424.1 318 Missense Mutation GCT,GTT A41V XP_016856913.1

View Full Product Details