Product Details
- SNP ID
-
rs140212391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:85062694 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAACGAGAGAGCAGTTAACGACAC[C/T]CTAAGAACTTTAAGGGCATGTACTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR63
PubMed Links
Gene Details
- Gene
- WDR63
- Gene Name
- WD repeat domain 63
View Full Product Details