Product Details

SNP ID
rs140231308
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109606072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTGGATCCACTTCTTTCTCTC[A/G]GATCTCTGCCCTCCCACATCAAACA
Phenotype
MIM: 139340 MIM: 611189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAT2 PubMed Links

Gene Details

Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005272.3 831 Silent Mutation TCC,TCT S206S NP_005263.1
XM_011541264.2 831 Silent Mutation TCC,TCT S206S XP_011539566.1
Gene
MIR197
Gene Name
microRNA 197
There are no transcripts associated with this gene.

View Full Product Details