Product Details

SNP ID
rs141021704
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155135941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGGCGGCTTTCTGGACTCGCT[C/T]ATTTACGGAGCATGCGTGGTCTTCA
Phenotype
MIM: 605951 MIM: 191164 MIM: 613683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPM3 PubMed Links

Gene Details

Gene
DPM3
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
There are no transcripts associated with this gene.

Gene
EFNA1
Gene Name
ephrin A1
There are no transcripts associated with this gene.

Gene
SLC50A1
Gene Name
solute carrier family 50 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122837.1 130 UTR 5 NP_001116309.1
NM_001122839.1 130 Silent Mutation CTC,CTT L10L NP_001116311.1
NM_001287586.1 130 UTR 5 NP_001274515.1
NM_001287587.1 130 UTR 5 NP_001274516.1
NM_001287588.1 130 UTR 5 NP_001274517.1
NM_001287589.1 130 UTR 5 NP_001274518.1
NM_001287590.1 130 Intron NP_001274519.1
NM_001287591.1 130 Intron NP_001274520.1
NM_001287592.1 130 Intron NP_001274521.1
NM_018845.3 130 Silent Mutation CTC,CTT L10L NP_061333.2
XM_005245339.4 130 Intron XP_005245396.1
XM_005245340.3 130 Intron XP_005245397.1
XM_005245341.4 130 Intron XP_005245398.1
XM_005245342.3 130 Intron XP_005245399.1
XM_006711453.2 130 Silent Mutation CTC,CTT L10L XP_006711516.1

View Full Product Details