Product Details

SNP ID
rs141615571
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:234631729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAGAATTTATTGTTGACTTGA[C/T]TACTAGCTCAAAGTAATCAGAAATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details