Product Details
- SNP ID
-
rs141712038
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:197510469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTTGGATGCAAGATTTAAGTTTG[C/G]TTTCCATTTGTGTTTTTGTCTGAAA
- Phenotype
-
MIM: 613292
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DENND1B
PubMed Links
Gene Details
- Gene
- DENND1B
- Gene Name
- DENN domain containing 1B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001195215.1 |
2131 |
Missense Mutation |
AAC,AAG |
N773K |
NP_001182144.1 |
NM_001195216.1 |
2131 |
Intron |
|
|
NP_001182145.1 |
NM_001300858.1 |
2131 |
Intron |
|
|
NP_001287787.1 |
NM_144977.4 |
2131 |
Intron |
|
|
NP_659414.2 |
XM_005244931.2 |
2131 |
Missense Mutation |
AAC,AAG |
N753K |
XP_005244988.1 |
XM_006711192.2 |
2131 |
Missense Mutation |
AAC,AAG |
N763K |
XP_006711255.1 |
XM_006711193.3 |
2131 |
Missense Mutation |
AAC,AAG |
N763K |
XP_006711256.1 |
XM_006711194.3 |
2131 |
Missense Mutation |
AAC,AAG |
N467K |
XP_006711257.1 |
XM_011509246.2 |
2131 |
Missense Mutation |
AAC,AAG |
N821K |
XP_011507548.1 |
XM_011509247.1 |
2131 |
Missense Mutation |
AAC,AAG |
N801K |
XP_011507549.1 |
XM_011509248.2 |
2131 |
Missense Mutation |
AAC,AAG |
N786K |
XP_011507550.1 |
XM_011509249.2 |
2131 |
Missense Mutation |
AAC,AAG |
N763K |
XP_011507551.1 |
XM_011509251.2 |
2131 |
Intron |
|
|
XP_011507553.1 |
XM_017000470.1 |
2131 |
Missense Mutation |
AAC,AAG |
N763K |
XP_016855959.1 |
XM_017000471.1 |
2131 |
Missense Mutation |
AAC,AAG |
N743K |
XP_016855960.1 |
XM_017000472.1 |
2131 |
Missense Mutation |
AAC,AAG |
N489K |
XP_016855961.1 |
View Full Product Details