Product Details
- SNP ID
-
rs141939942
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:225988266 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAAATTTGGCTGCCATCTCGACA[C/T]CATTGCTACCAATTTTTGGAGCATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDE2
PubMed Links
Gene Details
- Gene
- SDE2
- Gene Name
- SDE2 telomere maintenance homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152608.3 |
817 |
Missense Mutation |
GAT,GGT |
D255G |
NP_689821.3 |
View Full Product Details