Product Details

SNP ID
rs142698199
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153373863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTGGTAATGGGCAGCCTTCAG[C/T]GCAATGGCTACCAGGGATATGAATT
Phenotype
MIM: 603112
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A12 PubMed Links

Gene Details

Gene
S100A12
Gene Name
S100 calcium binding protein A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005621.1 311 Silent Mutation NP_005612.1

View Full Product Details