Product Details

SNP ID
rs143434276
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:182904427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCATAATTTCCTCTCTCTTTCCA[A/G]CTCCTGATTCATAGGGATAAAAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1625 Missense Mutation GCT,GTT A447V NP_112195.2

View Full Product Details