Product Details

SNP ID
rs143522244
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:159314324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCATGACTGTGAGCAGGAAGCA[G/T]TTGTTGATGCCAAAGGTGAGATAGA
Phenotype
MIM: 147140
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCER1A PubMed Links

Gene Details

Gene
FCER1A
Gene Name
Fc fragment of IgE receptor Ia
There are no transcripts associated with this gene.

Gene
OR10J3
Gene Name
olfactory receptor family 10 subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004467.1 336 Missense Mutation AAA,AAC K112N NP_001004467.1

View Full Product Details