Product Details

SNP ID
rs143766120
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:182904228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAATTGTCAAAGCAGCCCCTGT[A/G]AGAACACACACTCCTGTTCCTTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1824 Silent Mutation CTC,CTT L513L NP_112195.2

View Full Product Details