Product Details

SNP ID
rs145871831
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:158606831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGCCATCTGTGCTCCACTCCA[C/T]TATGCCAGCCACATGAATCCTACCC
Phenotype
MIM: 182860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10Z1 PubMed Links

Gene Details

Gene
OR10Z1
Gene Name
olfactory receptor family 10 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004478.1 393 Silent Mutation CAC,CAT H131H NP_001004478.1
Gene
SPTA1
Gene Name
spectrin alpha, erythrocytic 1
There are no transcripts associated with this gene.

View Full Product Details