Product Details

SNP ID
rs146583457
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:19307388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAGAGCTCCGTTTCCACCTGC[C/T]GGGTGGTGGCGTTGTACATGCCCTG
Phenotype
MIM: 603418 MIM: 614760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR7A2 PubMed Links

Gene Details

Gene
AKR7A2
Gene Name
aldo-keto reductase family 7 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320979.1 531 Missense Mutation CAG,CGG Q170R NP_001307908.1
NM_003689.3 531 Missense Mutation CAG,CGG Q205R NP_003680.2
Gene
PQLC2
Gene Name
PQ loop repeat containing 2
There are no transcripts associated with this gene.

View Full Product Details