Product Details

SNP ID
rs148447837
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:64474668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTACATTCTCGAGGAGGAGATA[A/C]GATTGCACAGGTAAACCAGCTAAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CACHD1 PubMed Links

Gene Details

Gene
CACHD1
Gene Name
cache domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293274.1 Intron NP_001280203.1
NM_020925.2 Intron NP_065976.2
XM_011541862.1 Intron XP_011540164.1
XM_011541863.1 Intron XP_011540165.1

View Full Product Details