Product Details

SNP ID
rs148863716
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:178851276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAGGTTAGAATGTGCACAGGC[A/G]TTGTACCACCAGCCTCCTTTATGAA
Phenotype
MIM: 603874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL1 PubMed Links

Gene Details

Gene
ANGPTL1
Gene Name
angiopoietin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004673.3 1805 Silent Mutation AAC,AAT N443N NP_004664.1
XM_005245577.2 1805 Silent Mutation AAC,AAT N443N XP_005245634.1
Gene
RALGPS2
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286247.1 1805 Intron NP_001273176.1
NM_152663.4 1805 Intron NP_689876.2
XM_006711410.3 1805 Intron XP_006711473.1
XM_006711411.3 1805 Intron XP_006711474.1
XM_011509688.1 1805 Intron XP_011507990.1
XM_017001591.1 1805 Intron XP_016857080.1

View Full Product Details