Product Details

SNP ID
rs148895280
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:203128872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCCATCTCAGCTTTCCAGGCC[G/T]CCTACATCGGCATCGAGGTGCTCAT
Phenotype
MIM: 102775
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 464 Missense Mutation GCC,TCC A11S NP_000665.1
NM_001048230.1 464 Missense Mutation GCC,TCC A11S NP_001041695.1
XM_005244901.1 464 UTR 5 XP_005244958.1
XM_005244902.4 464 Intron XP_005244959.1
XM_017000342.1 464 UTR 5 XP_016855831.1

View Full Product Details