Product Details

SNP ID
rs149563285
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:179082992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACCTGGACCTCCTGACCACGTG[A/G]TACTGCAGCTTCAAAGACTGCTGCC
Phenotype
MIM: 611063 MIM: 607555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM20B PubMed Links

Gene Details

Gene
FAM20B
Gene Name
family with sequence similarity 20 member B
There are no transcripts associated with this gene.

Gene
TOR3A
Gene Name
torsin family 3 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022371.3 464 Missense Mutation TGA,TGG *104W NP_071766.2

View Full Product Details