Product Details

SNP ID
rs149917617
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:229629185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTGTATAGATAGGCACAGTCCT[C/T]CTTTTCATAAAGCACTTGTATGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF5L PubMed Links

Gene Details

Gene
TAF5L
Gene Name
TATA-box binding protein associated factor 5 like
There are no transcripts associated with this gene.

Gene
URB2
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314021.1 Intron NP_001300950.1
NM_014777.3 Intron NP_055592.2
XM_005273360.3 Intron XP_005273417.1

View Full Product Details