Product Details

SNP ID
rs149959514
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152676312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGAGAGTGAACCTTCTGGGGG[A/C]TCTGGCTGCTGCCACAGCTCTGGGG
Phenotype
MIM: 612610 MIM: 612611
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCE2B PubMed Links

Gene Details

Gene
LCE2B
Gene Name
late cornified envelope 2B
There are no transcripts associated with this gene.

Gene
LCE2C
Gene Name
late cornified envelope 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178429.3 332 Silent Mutation GGA,GGC G99G NP_848516.1

View Full Product Details