Product Details

SNP ID
rs150447801
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:179087412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTGCCTCTGGGCACATGAGC[C/T]TCTCTTTGCCCATCACCCCTGTATG
Phenotype
MIM: 607555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOR3A PubMed Links

Gene Details

Gene
TOR3A
Gene Name
torsin family 3 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022371.3 Intron NP_071766.2

View Full Product Details