Product Details
- SNP ID
-
rs150666132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:19308469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCCCTCACCTCCTGGTGCAGCC[A/G]CTGGCAGGCATGCAGCGTCTCTTCC
- Phenotype
-
MIM: 603418
MIM: 614760
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKR7A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs859208] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKR7A2
- Gene Name
- aldo-keto reductase family 7 member A2
- Gene
- PQLC2
- Gene Name
- PQ loop repeat containing 2
There are no transcripts associated with this gene.
View Full Product Details