Product Details

SNP ID
rs143579230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23491871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGGACAAGTATGTCTTCCTGGT[C/G]GTCAAGACACTGCAAGCCCAGCTTC
Phenotype
MIM: 608683
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CST8 PubMed Links

Gene Details

Gene
CST8
Gene Name
cystatin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281730.1 589 Silent Mutation GTC,GTG V68V NP_001268659.1
NM_005492.3 589 Silent Mutation GTC,GTG V68V NP_005483.1

View Full Product Details