Product Details

SNP ID
rs146325379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63708883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCGGCCCGTGGGTCCGGATC[A/G]GAGACCGTTCCTAAAGCAGAGGCGG
Phenotype
MIM: 604699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFRP1 PubMed Links

Gene Details

Gene
ARFRP1
Gene Name
ADP ribosylation factor related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134758.3 473 Intron NP_001128230.1
NM_001267544.2 473 Intron NP_001254473.1
NM_001267545.2 473 Intron NP_001254474.1
NM_001267546.2 473 Intron NP_001254475.1
NM_001267547.2 473 Intron NP_001254476.1
NM_001267548.2 473 Intron NP_001254477.1
NM_001267549.2 473 Intron NP_001254478.1
NM_003224.5 473 Intron NP_003215.1
XM_011528482.2 473 Intron XP_011526784.1
XM_011528483.1 473 Intron XP_011526785.1
XM_017027576.1 473 Intron XP_016883065.1
Gene
ZGPAT
Gene Name
zinc finger CCCH-type and G-patch domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083113.1 473 Silent Mutation TCA,TCG S101S NP_001076582.1
NM_001195653.1 473 Silent Mutation TCA,TCG S101S NP_001182582.1
NM_001195654.1 473 Silent Mutation TCA,TCG S101S NP_001182583.1
NM_032527.4 473 Silent Mutation TCA,TCG S101S NP_115916.3
NM_181485.2 473 Silent Mutation TCA,TCG S101S NP_852150.2

View Full Product Details