Product Details

SNP ID
rs149653661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45650548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCCAGCACAGACAGTAGCACC[A/G]TACAGAAGAATGTCATGAGCATGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WFDC11 PubMed Links

Gene Details

Gene
WFDC11
Gene Name
WAP four-disulfide core domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147197.2 248 Missense Mutation NP_671730.1

View Full Product Details