Product Details

SNP ID
rs138899581
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45984391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGCGACGCACTCAAAAGCAGCG[C/T]GGACGCGGTCAAGTACTTTGGGAAG
Phenotype
MIM: 120220
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
COL6A1 PubMed Links
Additional Information
For this assay, SNP(s) [rs11553519] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
COL6A1
Gene Name
collagen type VI alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001848.2 452 Missense Mutation GCG,GTG A117V NP_001839.2
Gene
LOC101928796
Gene Name
uncharacterized LOC101928796
There are no transcripts associated with this gene.

View Full Product Details