Product Details

SNP ID
rs139648899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45984450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGACTGCGCTATCAAGAAGGGG[C/G]TGGAGCAGCTCCTCGTGGGGTGAGT
Phenotype
MIM: 120220
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL6A1 PubMed Links

Gene Details

Gene
COL6A1
Gene Name
collagen type VI alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001848.2 511 Missense Mutation CTG,GTG L137V NP_001839.2
Gene
LOC101928796
Gene Name
uncharacterized LOC101928796
There are no transcripts associated with this gene.

View Full Product Details